Alfa Aesar™ 3-Methylbenzo[b]thiophene-2-carboxaldehyde, 98%

Manufacturer: Alfa Aesar™ A1137506

Quantity 5g

 View more versions of this product

Catalog No. AAA1137506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22053-74-3
    Molecular Formula C10H8OS
    Formula Weight 176.24
    MDL Number MFCD00052307
    Beilstein 121582
    Synonym 3-Methylthianaphthene-2-carboxaldehyde
    Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxaldehyde

    Specifications

    Formula Weight 176.24
    Molecular Formula C10H8OS
    Synonym 3-Methylthianaphthene-2-carboxaldehyde
    Melting Point 88° to 90°C
    Quantity 5g
    MDL Number MFCD00052307
    CAS 22053-74-3
    Beilstein 121582
    Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxaldehyde
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit