Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzo[b]thiophene-2-carboxylic acid, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
3133-78-6
C10H8O2S
192.232
MFCD01830305
YVKLUKXESFJRCE-UHFFFAOYSA-N
3-methylbenzo b thiophene-2-carboxylic acid,3-methylbenzothiophene-2-carboxylic acid,acmc-20a6oy,methylbenzothiophene-carboxylic,3-methylthianaphthene-2-carboxylic acid,3-methyl 2-benzo b thiophenecarboxylic acid,benzo b thiophene-2-carboxylicacid, 3-methyl
821868
3-methyl-1-benzothiophene-2-carboxylic acid
CC1=C(SC2=CC=CC=C12)C(=O)O
3133-78-6
C10H8O2S
192.232
MFCD01830305
YVKLUKXESFJRCE-UHFFFAOYSA-N
3-methylbenzo b thiophene-2-carboxylic acid,3-methylbenzothiophene-2-carboxylic acid,acmc-20a6oy,methylbenzothiophene-carboxylic,3-methylthianaphthene-2-carboxylic acid,3-methyl 2-benzo b thiophenecarboxylic acid,benzo b thiophene-2-carboxylicacid, 3-methyl
821868
3-methyl-1-benzothiophene-2-carboxylic acid
CC1=C(SC2=CC=CC=C12)C(=O)O
Quantity 1g
Melting Point 241°C to 244°C
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning