Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzo[f]quinoline, 97%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
85-06-3
C14H11N
193.249
MFCD00046906
SUHRSZJZYUCLOD-UHFFFAOYSA-N
3-methylbenzo f quinoline,benzo f quinoline, 3-methyl,beta-naphthoquinaldine,.beta.-naphthoquinaldine,unii-j67ocr12xn,j67ocr12xn
6797
3-methylbenzo[f]quinoline
CC1=NC2=C(C=C1)C3=CC=CC=C3C=C2
85-06-3
C14H11N
193.249
MFCD00046906
SUHRSZJZYUCLOD-UHFFFAOYSA-N
3-methylbenzo f quinoline,benzo f quinoline, 3-methyl,beta-naphthoquinaldine,.beta.-naphthoquinaldine,unii-j67ocr12xn,j67ocr12xn
6797
3-methylbenzo[f]quinoline
CC1=NC2=C(C=C1)C3=CC=CC=C3C=C2
Applications
3-Methylbenzo[f]quinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point 73°C to 78°C
Quantity 25g
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[f]quinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA