Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzofuran, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
21535-97-7
C9H8O
132.16
MFCD00205641
ZRXHLJNBNWVNIM-UHFFFAOYSA-N
3-methylbenzofuran, 3-methyl-benzofuran, benzofuran, 3-methyl, 3-methylbenzo b furan, acmc-209flj, #
88939
3-methyl-1-benzofuran
CC1=COC2=CC=CC=C12
21535-97-7
C9H8O
132.16
MFCD00205641
ZRXHLJNBNWVNIM-UHFFFAOYSA-N
3-methylbenzofuran, 3-methyl-benzofuran, benzofuran, 3-methyl, 3-methylbenzo b furan, acmc-209flj, #
88939
3-methyl-1-benzofuran
CC1=COC2=CC=CC=C12
Refractive Index 1.5535 to 1.5555
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 132.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methylbenzofuran
Show More Show Less

EINECSNumber : 244-427-2