Alfa Aesar™ 3-Methylbenzophenone, 97%

Manufacturer: Alfa Aesar™ B2566114

Quantity 25g

 View more versions of this product

Catalog No. AAB2566114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 643-65-2
    Molecular Formula C14H12O
    Formula Weight 196.25
    MDL Number MFCD00008535
    Beilstein 2045488
    Synonym Phenyl m-tolyl ketone
    Chemical Name or Material 3-Methylbenzophenone

    Specifications

    Formula Weight 196.25
    Beilstein 2045488
    Flash Point >110°C (230°F)
    Boiling Point 183° to 185°C (16mmHg)
    Molecular Formula C14H12O
    MDL Number MFCD00008535
    CAS 643-65-2
    Synonym Phenyl m-tolyl ketone
    Quantity 25g
    Chemical Name or Material 3-Methylbenzophenone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit