Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methylbenzyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1078436

Quantity 500g

 View more versions of this product

Catalog No. AAA1078436


  Check Availability
Add to cart

Specifications

Description

Applications
3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Description & Specifications

Chemical Identifiers

620-19-9
140.61
UN3265
14,10091
3-Methylbenzyl chloride
C8H9Cl
MFCD00000909
2040357
alpha-Chloro-m-xylene; m-Xylyl chloride

Specifications

620-19-9
140.61
UN3265
14,10091
195° to 196°C
75°C (167°F)
3-Methylbenzyl chloride
C8H9Cl
MFCD00000909
2040357
alpha-Chloro-m-xylene; m-Xylyl chloride
500g
Pungent
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit