Alfa Aesar™ 3-Methylbenzyl isothiocyanate, 99%

Manufacturer: Alfa Aesar™ H5489303

Quantity 1g

 View more versions of this product

Catalog No. AAH5489303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3696-66-0
    Formula Weight 163.24
    MDL Number MFCD00060410
    UN Number UN2922
    Chemical Name or Material 3-Methylbenzyl isothiocyanate

    Specifications

    Formula Weight 163.24
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.09g/mL
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    MDL Number MFCD00060410
    UN Number UN2922
    Color Pale brown
    Health Hazard 1 Danger
    Quantity 1g
    CAS 3696-66-0
    Boiling Point 127°C (2mmHg)
    Chemical Name or Material 3-Methylbenzyl isothiocyanate
    Assay 99%

    GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit