Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
29875-06-7,109-99-9
C8H9ClMg
164.92
MPIXEEVXDGICED-UHFFFAOYSA-M
3-methylbenzylmagnesium chloride solution,m-methylbenzylmagnesium chloride,3-methylbenzylmagnesium chloride,3-methylbenzyl magnesium chloride,ikenqfuqpclwft-uhfffaoysa-m,magnesium,chloro 3-methylphenyl methyl,3-methylbenzylmagnesium chloride, 0.5m in thf,3-methylbenzylmagnesium chloride, 0.25m in 2-methf,3-methylbenzylmagnesium chloride solution, 0.5 m in thf
14867646
magnesium;1-methanidyl-3-methylbenzene;chloride
CC1=CC=CC(=C1)[CH2-].[Mg+2].[Cl-]
29875-06-7,109-99-9
C8H9ClMg
164.92
MPIXEEVXDGICED-UHFFFAOYSA-M
3-methylbenzylmagnesium chloride solution,m-methylbenzylmagnesium chloride,3-methylbenzylmagnesium chloride,3-methylbenzyl magnesium chloride,ikenqfuqpclwft-uhfffaoysa-m,magnesium,chloro 3-methylphenyl methyl,3-methylbenzylmagnesium chloride, 0.5m in thf,3-methylbenzylmagnesium chloride, 0.25m in 2-methf,3-methylbenzylmagnesium chloride solution, 0.5 m in thf
14867646
magnesium;1-methanidyl-3-methylbenzene;chloride
CC1=CC=CC(=C1)[CH2-].[Mg+2].[Cl-]
Specific Gravity 0.948
Packaging AcroSeal™ Glass Bottle
Density 0.9480g/mL
Quantity 50mL
Color Yellow to Brown
Flash Point −17°C
CAS Min % 4.0,94.78
CAS Max % 5.22,96.0
Formula Weight 164.92
Physical Form Liquid
Chemical Name or Material 3-Methylbenzylmagnesium chloride
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
Suspected of causing cancer if inhaled.
Causes skin irritation.
Causes serious eye damage.
Harmful if swallowed.
Suspected of

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger