Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Methylbiphenyl-4-sulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
186551-47-3
C13H11ClO2S
266.75
OHVJVSGIYVAZFD-UHFFFAOYSA-N
3'-methyl-biphenyl-4-sulfonyl chloride,3'-methylbiphenyl-4-sulfonyl chloride,3'-methyl-1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 3'-methyl,4-3-methylphenyl benzenesulfonyl chloride,3/'-methylbiphenyl-4-sulfonyl chloride,3'-methyl 1,1'-biphenyl-4-sulfonyl chloride
10754271
4-(3-methylphenyl)benzenesulfonyl chloride
CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
186551-47-3
C13H11ClO2S
266.75
OHVJVSGIYVAZFD-UHFFFAOYSA-N
3'-methyl-biphenyl-4-sulfonyl chloride,3'-methylbiphenyl-4-sulfonyl chloride,3'-methyl-1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 3'-methyl,4-3-methylphenyl benzenesulfonyl chloride,3/'-methylbiphenyl-4-sulfonyl chloride,3'-methyl 1,1'-biphenyl-4-sulfonyl chloride
10754271
4-(3-methylphenyl)benzenesulfonyl chloride
CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 266.75
Percent Purity 97%
Chemical Name or Material 3'-Methylbiphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger