Alfa Aesar™ 3-Methylcyclohexane-1,2-dione, 98+%

Manufacturer: Alfa Aesar™ B2262314

Quantity 25g

 View more versions of this product

Catalog No. AAB2262314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3008-43-3
    Molecular Formula C7H10O2
    Formula Weight 126.16
    MDL Number MFCD00209518
    Chemical Name or Material 3-Methylcyclohexane-1,2-dione

    Specifications

    Formula Weight 126.16
    TSCA TSCA
    Melting Point 57° to 59°C
    Molecular Formula C7H10O2
    MDL Number MFCD00209518
    CAS 3008-43-3
    Boiling Point 69° to 72°C (1mmHg)
    Quantity 25g
    Chemical Name or Material 3-Methylcyclohexane-1,2-dione
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit