Alfa Aesar™ 3-Methylcyclopentane-1,2-dione, 98+%

Manufacturer: Alfa Aesar™ A1025522

Quantity 100g

 View more versions of this product

Catalog No. AAA1025522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 765-70-8
    Molecular Formula C6H8O2
    Formula Weight 112.13
    MDL Number MFCD00001417
    Beilstein 2076520
    Synonym 2-Hydroxy-3-methyl-2-cyclopenten-1-one
    Chemical Name or Material 3-Methylcyclopentane-1,2-dione

    Specifications

    Formula Weight 112.13
    CAS 765-70-8
    TSCA TSCA
    Molecular Formula C6H8O2
    MDL Number MFCD00001417
    Beilstein 2076520
    Synonym 2-Hydroxy-3-methyl-2-cyclopenten-1-one
    Melting Point 103° to 107°C
    Quantity 100g
    Chemical Name or Material 3-Methylcyclopentane-1,2-dione
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit