Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methylcytosine (3-mC) Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
$135.70 - $519.80
Specifications
Antigen | 3-Methylcytosine (3-mC) |
---|---|
Concentration | Conc. Not Determined |
Applications | Dot Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4144
|
Active Motif
61180 |
10 μL |
Each for $135.70
|
|
|||||
50-199-4143
|
Active Motif
61179 |
100 μL |
Each for $519.80
|
|
|||||
Description
3-methylcytosine arises by erroneous methylation of DNA through spontaneous exposure to endogenous S-adenosyl methionine (SAM). Since the N3-position in cytosine is protected in double stranded DNA, this phenomenon mainly occurs in single stranded DNA. The resulting 3-mC is toxic and mutagenic and increased levels of 3-mC seem to impair cell proliferation. Therefore, it has to be repaired by the cell. This is established by oxidative demethylation, which is catalyzed by the AlkB protein.Specifications
3-Methylcytosine (3-mC) | |
Dot Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
3mc; 3-mC; 3-meC; 3-Methyl-cytosine; C5H7NO3 | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles |
Conc. Not Determined | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
This 3-methylcytosine antibody was raised against 3-methylcytidine conjugated to kLH and recognizes 3-methylcytosine . | |
Primary | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title