Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-methylcytosine(3-mC) Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP262790
Description
3-methylcytosine(3-mC) Polyclonal antibody specifically detects 3-methylcytosine(3-mC) in Human samples. It is validated for Dot Blot.Specifications
3-methylcytosine(3-mC) | |
Unconjugated | |
Purified | |
3-mC, 3-Methylcytosine, 6-amino-1-methylpyrimidin-2-one | |
3-methylcytosine (3-mC) | |
Polyclonal | |
Protein G purified | |
Store at -20°C. Avoid freeze-thaw cycles. |
Dot Blot | |
Dot Blot 1:400 | |
PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
Rabbit | |
IgG | |
Primary | |
50μg | |
Human,Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title