Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyldiphenylamine, 98%, Acros Organics

Click to view available options
Quantity:
10g
1205-64-7
C13H13N
183.254
MFCD00008530
TWPMMLHBHPYSMT-UHFFFAOYSA-N
3-methyldiphenylamine,n-phenyl-m-toluidine,benzenamine, 3-methyl-n-phenyl,n-phenyl-3-methylaniline,3-methyl diphenylamine,3-methyl diphenyl amine,3-methyl-n-phenyl-aniline,n-3-methylphenyl-aniline,3-methylphenyl phenylamine,n-3-methylphenyl aniline
14569
3-methyl-N-phenylaniline
CC1=CC(=CC=C1)NC2=CC=CC=C2
1205-64-7
C13H13N
183.254
MFCD00008530
TWPMMLHBHPYSMT-UHFFFAOYSA-N
3-methyldiphenylamine,n-phenyl-m-toluidine,benzenamine, 3-methyl-n-phenyl,n-phenyl-3-methylaniline,3-methyl diphenylamine,3-methyl diphenyl amine,3-methyl-n-phenyl-aniline,n-3-methylphenyl-aniline,3-methylphenyl phenylamine,n-3-methylphenyl aniline
14569
3-methyl-N-phenylaniline
CC1=CC(=CC=C1)NC2=CC=CC=C2
Color Brown to Black
Refractive Index 1.6330 to 1.6350 (20°C, 589nm)
Quantity 10g
Infrared Spectrum Authentic
Density 1.06g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4NHC6H5
Beilstein 12, 857
Formula Weight 183.25
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material 3-Methyldiphenylamine, 98%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.