Alfa Aesar™ 3-Methyldiphenylamine, 99%

Manufacturer: Alfa Aesar™ B2544109

Quantity 10g

 View more versions of this product

Catalog No. AAB2544109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1205-64-7
    Molecular Formula C13H13N
    Formula Weight 183.25
    MDL Number MFCD00008530
    Synonym N-Phenyl-m-toluidine
    Chemical Name or Material 3-Methyldiphenylamine

    Specifications

    Formula Weight 183.25
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Flash Point 147°C (297°F)
    Melting Point 25° to 27°C
    Quantity 10g
    TSCA TSCA
    Molecular Formula C13H13N
    MDL Number MFCD00008530
    CAS 1205-64-7
    Synonym N-Phenyl-m-toluidine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Boiling Point 317°C
    Chemical Name or Material 3-Methyldiphenylamine
    Assay 99%

    GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit