Alfa Aesar™ 3-Methylglutaric acid, 95%

Manufacturer: Alfa Aesar™ B2473714

Quantity 25g

 View more versions of this product

Catalog No. AAB2473714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 626-51-7
    Molecular Formula C6H10O4
    Formula Weight 146.14
    MDL Number MFCD00002727
    Beilstein 1759502
    Synonym 3-Methylpropane-1,3-dicarboxylic acid; 3-Methylpentanedioic acid
    Chemical Name or Material 3-Methylglutaric acid

    Specifications

    Formula Weight 146.14
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 78° to 83°C
    Molecular Formula C6H10O4
    MDL Number MFCD00002727
    CAS 626-51-7
    Beilstein 1759502
    Synonym 3-Methylpropane-1,3-dicarboxylic acid; 3-Methylpentanedioic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 25g
    Chemical Name or Material 3-Methylglutaric acid
    Assay 95%

    Description

    Applications
    3-Methylglutaric acid is a hypolipidemic agent and also used as an chemical, organic intermediate, pharmaceutical intermediate.

    Solubility
    Soluble in water, and ethanol.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit