Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylglutaric Acid 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
626-51-7
C6H10O4
146.142
MFCD00002727
XJMMNTGIMDZPMU-UHFFFAOYSA-N
3-methylglutaric acid,methylglutaric acid,pentanedioic acid, 3-methyl,3-methyl-pentanedioic acid,3-methylglutaricacid,beta-methylglutaric acid,unii-3q0p190c7b,3-methylpentanedioate,b-methylglutarate,3-methylglutarate
12284
CHEBI:68566
3-methylpentanedioic acid
CC(CC(=O)O)CC(=O)O
626-51-7
C6H10O4
146.142
MFCD00002727
XJMMNTGIMDZPMU-UHFFFAOYSA-N
3-methylglutaric acid,methylglutaric acid,pentanedioic acid, 3-methyl,3-methyl-pentanedioic acid,3-methylglutaricacid,beta-methylglutaric acid,unii-3q0p190c7b,3-methylpentanedioate,b-methylglutarate,3-methylglutarate
12284
CHEBI:68566
3-methylpentanedioic acid
CC(CC(=O)O)CC(=O)O
Melting Point 83°C to 87°C
Color White
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HO2CCH2CH(CH3)CH2CO2H
Beilstein 02, 659
Formula Weight 146.14
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 3-Methylglutaric acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.