Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylhexanoic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
3780-58-3
C7H14O2
130.19
NZQMQVJXSRMTCJ-UHFFFAOYSA-N
3-methylcaproic acid,3-methyl-hexanoic acid,hexanoic acid, 3-methyl,3-methylhexylic acid,3-methyl hexanoic acid,hexanoicacid,3-methyl,3-methyl-2-hexanoic acid,2-02-00-00298 beilstein handbook reference
95315
3-methylhexanoic acid
CCCC(C)CC(=O)O
3780-58-3
C7H14O2
130.19
NZQMQVJXSRMTCJ-UHFFFAOYSA-N
3-methylcaproic acid,3-methyl-hexanoic acid,hexanoic acid, 3-methyl,3-methylhexylic acid,3-methyl hexanoic acid,hexanoicacid,3-methyl,3-methyl-2-hexanoic acid,2-02-00-00298 beilstein handbook reference
95315
3-methylhexanoic acid
CCCC(C)CC(=O)O
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Boiling Point 215.0°C to 216.0°C
Flash Point 102°C
Assay Percent Range 96.0 to max. 104% min. (Acid)
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 3-Methylhexanoic acid

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger