Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methylindole-2-carboxaldehyde, 97%

Click to view available options
Quantity:
250mg
1g
5257-24-9
C10H9NO
159.188
MFCD00957066
GHSUORIVDBPKKQ-UHFFFAOYSA-N
3-methylindole-2-carbaldehyde,3-methyl-1h-indole-2-carboxaldehyde,3-methylindole-2-carboxaldehyde,1h-indole-2-carboxaldehyde, 3-methyl,methylindolaldehyd,3-methyl-2-indolecarbaldehyde,3-methyl-indole-2-carbaldehyde,2-formyl-3-methylindole,3-methyl-2-hydroxymethylenindol,3-methyl-1h-indole-2-carbaldehyd
277260
3-methyl-1H-indole-2-carbaldehyde
CC1=C(NC2=CC=CC=C12)C=O
5257-24-9
C10H9NO
159.188
MFCD00957066
GHSUORIVDBPKKQ-UHFFFAOYSA-N
3-methylindole-2-carbaldehyde,3-methyl-1h-indole-2-carboxaldehyde,3-methylindole-2-carboxaldehyde,1h-indole-2-carboxaldehyde, 3-methyl,methylindolaldehyd,3-methyl-2-indolecarbaldehyde,3-methyl-indole-2-carbaldehyde,2-formyl-3-methylindole,3-methyl-2-hydroxymethylenindol,3-methyl-1h-indole-2-carbaldehyd
277260
3-methyl-1H-indole-2-carbaldehyde
CC1=C(NC2=CC=CC=C12)C=O
Sensitivity Air & Moisture Sensitive
Quantity 250mg
Beilstein 4034
Formula Weight 159.18
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Methylindole-2-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning