Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylindole 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
Plastic bottle
83-34-1
C9H9N
131.18
MFCD00005627
ZFRKQXVRDFCRJG-UHFFFAOYSA-N
3-methylindole,skatole,scatole,skatol,1h-indole, 3-methyl,beta-methylindole,indole, 3-methyl,3-mi,3-methyl-4,5-benzopyrrole,3-methyl indole
6736
CHEBI:9171
3-methyl-1H-indole
CC1=CNC2=CC=CC=C12
83-34-1
C9H9N
131.18
MFCD00005627
ZFRKQXVRDFCRJG-UHFFFAOYSA-N
3-methylindole,skatole,scatole,skatol,1h-indole, 3-methyl,beta-methylindole,indole, 3-methyl,3-mi,3-methyl-4,5-benzopyrrole,3-methyl indole
6736
CHEBI:9171
3-methyl-1H-indole
CC1=CNC2=CC=CC=C12
Flash Point 132°C
Packaging Plastic bottle
Melting Point 95.0°C to 98.0°C
Quantity 25g
Color Brown to White
Boiling Point 265.0°C to 266.0°C (755.0mmHg)
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 20, 315
Merck Index 15, 8698
Solubility Information Solubility in water: 0.45g/L (20°C). Other solubilities: soluble in hot water,soluble in alcohol,benzene,chloroform and ether
Formula Weight 131.18
Physical Form Crystalline Powder or Flakes
Percent Purity 98%
Chemical Name or Material 3-Methylindole, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-471-7

RTECSNumber : NM0350000

TSCA : TSCA