Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylisoxazole-5-carboxylic acid, 97%, Acros Organics™

Click to view available options
Quantity:
5g
4857-42-5
C5H5NO3
127.1
HXIYCKAAQPHZBM-UHFFFAOYSA-N
3-methylisoxazole-5-carboxylic acid,3-methyl-5-isoxazolecarboxylic acid,5-isoxazolecarboxylic acid, 3-methyl,3-methyl-isoxazole-5-carboxylic acid,5-carboxy-3-methylisoxazole,5-isoxazolecarboxylicacid, 3-methyl,3-methylisoxazole-5-carboxylicacid,acmc-1aruu,3-methyl-5-carboxyisoxazole,aronis24519
853085
3-methyl-1,2-oxazole-5-carboxylic acid
CC1=NOC(=C1)C(=O)O
4857-42-5
C5H5NO3
127.1
HXIYCKAAQPHZBM-UHFFFAOYSA-N
3-methylisoxazole-5-carboxylic acid,3-methyl-5-isoxazolecarboxylic acid,5-isoxazolecarboxylic acid, 3-methyl,3-methyl-isoxazole-5-carboxylic acid,5-carboxy-3-methylisoxazole,5-isoxazolecarboxylicacid, 3-methyl,3-methylisoxazole-5-carboxylicacid,acmc-1aruu,3-methyl-5-carboxyisoxazole,aronis24519
853085
3-methyl-1,2-oxazole-5-carboxylic acid
CC1=NOC(=C1)C(=O)O
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 3-Methylisoxazole-5-carboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 225-454-9