Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyloxetane-3-carboxylic acid, 96%

Click to view available options
Quantity:
250mg
1g
28562-68-7
C5H8O3
116.116
MFCD09842435
PMPZEEUNGQSAIQ-UHFFFAOYSA-N
3-methyloxetane-3-carboxylicacid,3-methyl-3-oxetanecarboxylic acid,3-oxetanecarboxylic acid, 3-methyl,3-methyl-oxetane-3-carboxylic acid,pubchem14214,ksc199o1l,3-methyl oxetane-3-carboxylic acid,3-methyl-3-oxetane carboxylic acid,3-oxetanecarboxylicacid, 3-methyl,3-methyloxetane-3-carboxylic acid
14028997
3-methyloxetane-3-carboxylic acid
CC1(COC1)C(=O)O
28562-68-7
C5H8O3
116.116
MFCD09842435
PMPZEEUNGQSAIQ-UHFFFAOYSA-N
3-methyloxetane-3-carboxylicacid,3-methyl-3-oxetanecarboxylic acid,3-oxetanecarboxylic acid, 3-methyl,3-methyl-oxetane-3-carboxylic acid,pubchem14214,ksc199o1l,3-methyl oxetane-3-carboxylic acid,3-methyl-3-oxetane carboxylic acid,3-oxetanecarboxylicacid, 3-methyl,3-methyloxetane-3-carboxylic acid
14028997
3-methyloxetane-3-carboxylic acid
CC1(COC1)C(=O)O
Sensitivity Moisture Sensitive
Color White to Yellow
Quantity 1g
Melting Point 58°C to 63°C
Formula Weight 116.12
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 3-Methyloxetane-3-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C