Alfa Aesar™ 3-Methyloxindole, 97%

Manufacturer: Alfa Aesar™ H2766214

Quantity 25g

 View more versions of this product

Catalog No. AAH2766214

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1504-06-9
    Molecular Formula C9H9NO
    Formula Weight 147.18
    MDL Number MFCD04037370
    Chemical Name or Material 3-Methyloxindole

    Specifications

    Formula Weight 147.18
    MDL Number MFCD04037370
    CAS 1504-06-9
    Melting Point 121° to 124°C
    Quantity 25g
    Molecular Formula C9H9NO
    Chemical Name or Material 3-Methyloxindole
    Assay 97%

    Description

    Applications
    A versatile reactant. As 3-Methylindole metabolite. Reactant for enantioselective α-amination reactions, Reactant for aldol reaction with glyoxal derivatives, Reactant for amine thiourea catalyzed conjugate addition to α,β-unsaturated aldehydes, Reactant for O-acetylation reactions, Reactant for preparation of a disubstituted oxoindole by using rhodium-catalyzed cyclopropanation/ring-opening reactions.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit