Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpentyl Acetate 98.0+%, TCI America™

Click to view available options
25ML
35897-13-3
C8H16O2
144.214
MFCD00671487
NFYJZIICTJCGOR-UHFFFAOYSA-N
Acetic Acid 3-Methylpentyl Ester
3015816
3-methylpentyl acetate
CCC(C)CCOC(=O)C
35897-13-3
C8H16O2
144.214
MFCD00671487
NFYJZIICTJCGOR-UHFFFAOYSA-N
Acetic Acid 3-Methylpentyl Ester
3015816
3-methylpentyl acetate
CCC(C)CCOC(=O)C
Boiling Point 168°C
Color Colorless
Quantity 25mL
UN Number 3272
Formula Weight 144.21
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Methylpentyl Acetate

EINECSNumber : (2)-0734

TSCA : No