Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylphenethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
55755-17-4
C9H13N
135.21
MFCD01310828
GUERDLPJJJMIEU-UHFFFAOYSA-N
3-methylphenethylamine,2-m-tolyl ethanamine,2-m-tolyl-ethylamine,2-m-tolylethanamine,2-3-methylphenyl ethanamine,2-3-methylphenyl ethylamine,2-3-methylphenyl ethan-1-amine,2-m-tolyl ethylamine,3,4-methyl-phenethylamine,2-m-tolyl ethan-1-amine
410085
2-(3-methylphenyl)ethanamine
CC1=CC(=CC=C1)CCN
55755-17-4
C9H13N
135.21
MFCD01310828
GUERDLPJJJMIEU-UHFFFAOYSA-N
3-methylphenethylamine,2-m-tolyl ethanamine,2-m-tolyl-ethylamine,2-m-tolylethanamine,2-3-methylphenyl ethanamine,2-3-methylphenyl ethylamine,2-3-methylphenyl ethan-1-amine,2-m-tolyl ethylamine,3,4-methyl-phenethylamine,2-m-tolyl ethan-1-amine
410085
2-(3-methylphenyl)ethanamine
CC1=CC(=CC=C1)CCN
Refractive Index 1.5255 to 1.5275
Packaging Glass bottle
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 12, IV, 2639
Formula Weight 135.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methylphenethylamine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA