Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methylphenoxyacetic acid, 98%

Manufacturer:  Alfa Aesar™ B2486922

Quantity 100g

 View more versions of this product

Catalog No. AAB2486922


  Check Availability
Add to cart

Specifications

Description

Applications
3-Methylphenoxyacetic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 4810 mg/L at 25°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Description & Specifications

Chemical Identifiers

1643-15-8
166.18
2047572
3-Methylphenoxyacetic acid
C9H10O3
MFCD00014358
m-Cresoxyacetic acid

Specifications

1643-15-8
166.18
2047572
102° to 105°C
3-Methylphenoxyacetic acid
C9H10O3
MFCD00014358
m-Cresoxyacetic acid
100g
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AI9316500

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit