Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpyridazine 99%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
1632-76-4
C5H6N2
94.12
MFCD00006469
MXDRPNGTQDRKQM-UHFFFAOYSA-N
pyridazine, 3-methyl,methyldiazine,pyridazine, methyl,methylpyridazine,3-methyl pyridazine,pubchem17696,3-methylpyridazin,acmc-1bp1x,3-methylpyridazine,ksc181a2j
74208
3-methylpyridazine
CC1=NN=CC=C1
1632-76-4
C5H6N2
94.12
MFCD00006469
MXDRPNGTQDRKQM-UHFFFAOYSA-N
pyridazine, 3-methyl,methyldiazine,pyridazine, methyl,methylpyridazine,3-methyl pyridazine,pubchem17696,3-methylpyridazin,acmc-1bp1x,3-methylpyridazine,ksc181a2j
74208
3-methylpyridazine
CC1=NN=CC=C1
Boiling Point 214°C
Flash Point 87°C
Specific Gravity 1.031
Refractive Index 1.5140 to 1.516
Packaging Glass bottle
Quantity 10g
Infrared Spectrum Authentic
Color Yellow to Brown
Density 1.0310g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23, 92
Formula Weight 94.12
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3-Methylpyridazine, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-640-0

TSCA : TSCA