Alfa Aesar™ 3-Methylthio-1-propanol, 98%

Manufacturer: Alfa Aesar™ A1040322

Quantity 100g

 View more versions of this product

Catalog No. AAA1040322

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 505-10-2
    Molecular Formula CH3S(CH2)3OH
    Formula Weight 106.19
    MDL Number MFCD00036560
    Synonym Methionol; 3-(Methylmercapto)propan-1-ol
    Chemical Name or Material 3-Methylthio-1-propanol

    Specifications

    Formula Weight 106.19
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Methionol; 3-(Methylmercapto)propan-1-ol
    Quantity 100g
    TSCA TSCA
    Molecular Formula CH3S(CH2)3OH
    CAS 505-10-2
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Boiling Point 89° to 90°C (13mmHg)
    MDL Number MFCD00036560
    Health Hazard 1 Warning
    Flash Point 90°C (195°F)
    Chemical Name or Material 3-Methylthio-1-propanol
    Assay 98%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit