Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthio-1-propanol, 98%, Thermo Scientific Chemicals

Catalog Number p-7080743
Click to view available options
Quantity:
5 g
25 g
100 g
505-10-2
C4H10OS
106.18
MFCD00036560
CZUGFKJYCPYHHV-UHFFFAOYSA-N
methionol, 3-methylthiopropanol, 3-methylthio-1-propanol, 3-hydroxypropyl methyl sulfide, 3-methylthio propanol, 3-methylmercapto-1-propanol, 1-propanol, 3-methylthio, 3-methylsulfanyl propan-1-ol, gamma-methylmercaptopropyl alcohol
10448
CHEBI:49019
3-methylsulfanylpropan-1-ol
CSCCCO
505-10-2
C4H10OS
106.18
MFCD00036560
CZUGFKJYCPYHHV-UHFFFAOYSA-N
methionol, 3-methylthiopropanol, 3-methylthio-1-propanol, 3-hydroxypropyl methyl sulfide, 3-methylthio propanol, 3-methylmercapto-1-propanol, 1-propanol, 3-methylthio, 3-methylsulfanyl propan-1-ol, gamma-methylmercaptopropyl alcohol
10448
CHEBI:49019
3-methylsulfanylpropan-1-ol
CSCCCO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.027
Boiling Point 89°C to 90°C (13 mmHg)
Flash Point 90°C (195°F)
Refractive Index 1.4904
Quantity 5 g
Formula Weight 106.19
Percent Purity 98%
Chemical Name or Material 3-Methylthio-1-propanol
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 208-004-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit