Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Methylthio)aniline, 97%

Click to view available options
Quantity:
5g
25g
100g
1783-81-9
C7H9NS
139.216
MFCD00007793
KCHLDNLIJVSRPK-UHFFFAOYSA-N
3-methylthio aniline,3-aminothioanisole,m-aminothioanisole,m-methylthio aniline,benzenamine, 3-methylthio,3-methylmercapto aniline,3-methylsulfanyl aniline,3-methylmercaptoaniline,3-methylthiobenzenamine,m-aminophenyl methyl sulfide
15693
3-methylsulfanylaniline
CSC1=CC=CC(=C1)N
1783-81-9
C7H9NS
139.216
MFCD00007793
KCHLDNLIJVSRPK-UHFFFAOYSA-N
3-methylthio aniline,3-aminothioanisole,m-aminothioanisole,m-methylthio aniline,benzenamine, 3-methylthio,3-methylmercapto aniline,3-methylsulfanyl aniline,3-methylmercaptoaniline,3-methylthiobenzenamine,m-aminophenyl methyl sulfide
15693
3-methylsulfanylaniline
CSC1=CC=CC(=C1)N
Boiling Point 163°C to 165°C (16mmHg)
Odor Unpleasant
Flash Point >110°C (230°F)
Quantity 100g
UN Number UN2810
Beilstein 2078599
Formula Weight 139.23
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)aniline

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : BY6301000

TSCA : TSCA