Learn More
3-(Methylthio)propionaldehyde, 97%, Thermo Scientific Chemicals
Description
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents
Solubility
It is readily soluble in alcohol solvents such as ethanol, propylene and glycol oil. It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases.
Specifications
Melting Point | -58°C |
Density | 1.052 |
Boiling Point | 165°C to 166°C |
Flash Point | 64°C (147°F) |
Odor | Strong |
Refractive Index | 1.4825 |
Linear Formula | CH3S(CH2)2CHO |
Quantity | 10 g |
UN Number | UN2785 |
Beilstein | 1739289 |
Show More |
Safety and Handling
Hazard Category | H227-H302+H332-H311-H315-H317-H318 |
Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H317-H335-H227 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P210-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P361-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 4-THIAPENTANAL |
EINECSNumber | 221-882-5 |
RTECSNumber | UE2285000 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.