Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Methylthio)propyl isothiocyanate, 98%

Click to view available options
Quantity:
10g
50g
505-79-3
C5H9NS2
147.254
MFCD00209521
LDKSCZJUIURGMW-UHFFFAOYSA-N
3-methylthio propyl isothiocyanate,iberverin,3-methylthiopropyl isothiocyanate,ibervirin,propane, 1-isothiocyanato-3-methylthio,unii-ytt13bn94z,3-methylmercaptopropyl isothiocyanate,isothiocyanic acid, 3-methylthio propyl ester,1-isothiocyanato-3-methylthio propane,fema no. 3312
62351
1-isothiocyanato-3-methylsulfanylpropane
CSCCCN=C=S
505-79-3
C5H9NS2
147.254
MFCD00209521
LDKSCZJUIURGMW-UHFFFAOYSA-N
3-methylthio propyl isothiocyanate,iberverin,3-methylthiopropyl isothiocyanate,ibervirin,propane, 1-isothiocyanato-3-methylthio,unii-ytt13bn94z,3-methylmercaptopropyl isothiocyanate,isothiocyanic acid, 3-methylthio propyl ester,1-isothiocyanato-3-methylthio propane,fema no. 3312
62351
1-isothiocyanato-3-methylsulfanylpropane
CSCCCN=C=S
Odor Pungent
Quantity 50g
Flash Point 110°C (230°F)
Boiling Point 254°C
UN Number UN2922
Formula Weight 147.25
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propyl isothiocyanate

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA