Alfa Aesar™ 3-Methylvaleryl chloride, 97%

Manufacturer: Alfa Aesar™ H5041503

Quantity 1g

 View more versions of this product

Catalog No. AAH5041503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51116-72-4
    Molecular Formula C6H11ClO
    Formula Weight 134.61
    MDL Number MFCD00039315
    UN Number UN2920
    Synonym 3-Methylpentanoyl chloride
    Chemical Name or Material 3-Methylvaleryl chloride

    Specifications

    Formula Weight 134.61
    Molecular Formula C6H11ClO
    MDL Number MFCD00039315
    UN Number UN2920
    Synonym 3-Methylpentanoyl chloride
    Health Hazard 1 Danger
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    CAS 51116-72-4
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
    Chemical Name or Material 3-Methylvaleryl chloride
    Assay 97%

    Description

    Applications
    3-Methylvaleryl chloride can react with methylbenzene to product 4-methyl-1-p-tolyl-pentan-1-one. This reaction needs reagent AlCl3.

    Solubility
    Reacts with water.

    Notes
    Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit