Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylxanthine, 97%, Acros Organics

Click to view available options
Quantity:
100mg
250mg
1g
1076-22-8
C6H6N4O2
166.14
GMSNIKWWOQHZGF-UHFFFAOYSA-N
3-methylxanthine,3-methyl-1h-purine-2,6 3h,7h-dione,2,6-dihydroxy-3-methylpurine,xanthine, 3-methyl,3 mx,3-methyl xanthine,1h-purine-2,6-dione, 3,7-dihydro-3-methyl,unii-ws6x982oec,3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione,ccris 5817
70639
CHEBI:62207
3-methyl-7H-purine-2,6-dione
CN1C2=C(C(=O)NC1=O)NC=N2
1076-22-8
C6H6N4O2
166.14
GMSNIKWWOQHZGF-UHFFFAOYSA-N
3-methylxanthine,3-methyl-1h-purine-2,6 3h,7h-dione,2,6-dihydroxy-3-methylpurine,xanthine, 3-methyl,3 mx,3-methyl xanthine,1h-purine-2,6-dione, 3,7-dihydro-3-methyl,unii-ws6x982oec,3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione,ccris 5817
70639
CHEBI:62207
3-methyl-7H-purine-2,6-dione
CN1C2=C(C(=O)NC1=O)NC=N2
Melting Point >300.0°C
Quantity 100mg
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Beilstein 26,453
Solubility Information Solubility in water: insoluble
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 3-Methylxanthine

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 214-058-1