Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methylxanthine, 97%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
250 mg
1 g
Specifications
Melting Point | >300.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 250 mg |
Beilstein | 26,453 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 166.14 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methylxanthine |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. |
EINECSNumber | 214-058-1 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction