Learn More
3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) Mouse anti-All, Clone: 6H, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS216925UL
Description
3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) Monoclonal antibody specifically detects 3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) in All samples. It is validated for ELISA.Specifications
3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
All species | |
Purified |
ELISA | |
6H | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
KLH-conjugate of 3-Monoglucuronyl Glycyrrhetinic acid (3-MGA). | |
25 μL | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |