Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) Mouse anti-All, Clone: 6H, MilliporeSigma™

Mouse Monoclonal Antibody

$387.00 - $391.00

Specifications

Antigen 3-Monoglucuronyl Glycyrrhetinic acid (3-MGA)
Clone 6H
Applications ELISA
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS2169100
View Documents
MilliporeSigma
MABS2169100UL
100 μL
Each for $387.00
Add to cart
 
MABS216925
View Documents
MilliporeSigma
MABS216925UL
25 μL
Each for $391.00
Add to cart
 
Description

Description

3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) Monoclonal antibody specifically detects 3-Monoglucuronyl Glycyrrhetinic acid (3-MGA) in All samples. It is validated for ELISA.
Specifications

Specifications

3-Monoglucuronyl Glycyrrhetinic acid (3-MGA)
ELISA
Purified
RUO
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
6H
Unconjugated
Mouse
All
KLH-conjugate of 3-Monoglucuronyl Glycyrrhetinic acid (3-MGA).
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit