Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Morpholinophenyl)methanol, 97%, Acros Organics

Click to view available options
Quantity:
5g
145127-38-4
C11H15NO2
193.24
MXBVALXTJZMIJB-UHFFFAOYSA-N
3-morpholinophenyl methanol,3-morpholinobenzyl alcohol,3-morpholin-4-yl phenyl methanol,3-morpholin-4-ylphenyl methanol,benzenemethanol,3-4-morpholinyl,benzenemethanol, 3-4-morpholinyl,3-morpholinobenzylalcohol,acmc-20n4f1,3-morpholin-4-ylphenyl methan-1-ol
7162074
(3-morpholin-4-ylphenyl)methanol
C1COCCN1C2=CC=CC(=C2)CO
145127-38-4
C11H15NO2
193.24
MXBVALXTJZMIJB-UHFFFAOYSA-N
3-morpholinophenyl methanol,3-morpholinobenzyl alcohol,3-morpholin-4-yl phenyl methanol,3-morpholin-4-ylphenyl methanol,benzenemethanol,3-4-morpholinyl,benzenemethanol, 3-4-morpholinyl,3-morpholinobenzylalcohol,acmc-20n4f1,3-morpholin-4-ylphenyl methan-1-ol
7162074
(3-morpholin-4-ylphenyl)methanol
C1COCCN1C2=CC=CC(=C2)CO
Quantity 5g
Melting Point 51.0°C to 56.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (3-Morpholinophenyl)methanol

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning