Alfa Aesar™ 3-n-Butyl-2,4-pentanedione, 98%

Manufacturer: Alfa Aesar™ A1123604

Quantity 2g

 View more versions of this product

Catalog No. AAA1123604

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1540-36-9
    Molecular Formula C9H16O2
    Formula Weight 156.23
    MDL Number MFCD00026235
    Beilstein 507436
    Synonym 3-Acetyl-2-heptanone
    Chemical Name or Material 3-n-Butyl-2,4-pentanedione

    Specifications

    Formula Weight 156.23
    Molecular Formula C9H16O2
    MDL Number MFCD00026235
    Boiling Point 101° to 104°C (10mmHg)
    Beilstein 507436
    Quantity 2g
    Synonym 3-Acetyl-2-heptanone
    CAS 1540-36-9
    Chemical Name or Material 3-n-Butyl-2,4-pentanedione
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit