Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-n-Hexylpyridine, 98+%

Click to view available options
Quantity:
2.5g
10g
6311-92-8
C11H17N
163.264
MFCD02683071
LWEQIZSFZFUWIE-UHFFFAOYSA-N
3-n-hexylpyridine,pyridine, 3-hexyl,3-n-hexylpyridine,98+%
22766
3-hexylpyridine
CCCCCCC1=CN=CC=C1
6311-92-8
C11H17N
163.264
MFCD02683071
LWEQIZSFZFUWIE-UHFFFAOYSA-N
3-n-hexylpyridine,pyridine, 3-hexyl,3-n-hexylpyridine,98+%
22766
3-hexylpyridine
CCCCCCC1=CN=CC=C1
Quantity 10g
Formula Weight 163.27
Percent Purity ≥98%
Chemical Name or Material 3-n-Hexylpyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning