Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-n-Hexylthio-1,2,4-triazole, 97%

Click to view available options
Quantity:
2.5g
10g
74682-60-3
C8H15N3S
185.289
MFCD02683096
NZXRSXPNMMTVHB-UHFFFAOYSA-N
3-hexylthio-1h-1,2,4-triazole,3-n-hexylthio-1,2,4-triazole,3-hexylthio-1,2,4-triazole,1h-1,2,4-triazole,5-hexylthio,3-hexylsulfanyl-1h-1,2,4-triazole,3-hexylsulfanyl-2h-1,2,4-triazole,3-hexylsulfanyl-4h-1,2,4-triazole
2818229
5-hexylsulfanyl-1H-1,2,4-triazole
CCCCCCSC1=NC=NN1
74682-60-3
C8H15N3S
185.289
MFCD02683096
NZXRSXPNMMTVHB-UHFFFAOYSA-N
3-hexylthio-1h-1,2,4-triazole,3-n-hexylthio-1,2,4-triazole,3-hexylthio-1,2,4-triazole,1h-1,2,4-triazole,5-hexylthio,3-hexylsulfanyl-1h-1,2,4-triazole,3-hexylsulfanyl-2h-1,2,4-triazole,3-hexylsulfanyl-4h-1,2,4-triazole
2818229
5-hexylsulfanyl-1H-1,2,4-triazole
CCCCCCSC1=NC=NN1
Melting Point 65°C to 68°C
Quantity 2.5g
Formula Weight 185.29
Percent Purity 97%
Chemical Name or Material 3-n-Hexylthio-1,2,4-triazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning