Alfa Aesar™ 3-n-Hexylthiophene, 99+%

Manufacturer: Alfa Aesar™ H5589106

Quantity 5g

 View more versions of this product

Catalog No. AAH5589106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1693-86-3
    Molecular Formula C10H18S
    Formula Weight 168.3
    MDL Number MFCD00143181
    UN Number UN1993
    Beilstein 1617129
    Chemical Name or Material 3-n-Hexylthiophene

    Specifications

    Formula Weight 168.3
    UN Number UN1993
    Odor Stench
    Quantity 5g
    Molecular Formula C10H18S
    MDL Number MFCD00143181
    CAS 1693-86-3
    Beilstein 1617129
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H226-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 37°C (99°F)
    Boiling Point 65°C (0.45mmHg)
    Chemical Name or Material 3-n-Hexylthiophene
    Assay ≥99%

    Description

    Applications
    3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

    Solubility
    Partly soluble in water.

    Notes
    Store at room temperature. Avoid strong oxidizing agents.

    GHS H Statement
    H301-H311-H226-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit