Learn More
Alfa Aesar™ 3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester, 97+%
Manufacturer:  Alfa Aesar™ H2794906
Quantity 5g
 View more versions of this product
Catalog No. AAH2794906
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
957061-17-5
291.16
[3-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate
C15H22BNO4
MFCD09258775
3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester
Specifications
957061-17-5
5g
291.16
[3-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate
≥97%
69° to 72°C
C15H22BNO4
MFCD09258775
3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester