Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine 96.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
172478-00-1
C10H20N2O2
200.282
MFCD00191665
XYKYUXYNQDXZTD-UHFFFAOYSA-N
3-(N-Boc-N-methylamino)pyrrolidine
2761404
tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate
CC(C)(C)OC(=O)N(C)C1CCNC1
172478-00-1
C10H20N2O2
200.282
MFCD00191665
XYKYUXYNQDXZTD-UHFFFAOYSA-N
3-(N-Boc-N-methylamino)pyrrolidine
2761404
tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate
CC(C)(C)OC(=O)N(C)C1CCNC1
Color Colorless
Boiling Point 96°C
Quantity 25g
Formula Weight 200.28
Physical Form Clear Liquid at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material 3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine