Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-1H-pyrazole, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
26621-44-3
C3H3N3O2
113.08
MZRUFMBFIKGOAL-UHFFFAOYSA-N
3-nitro-1h-pyrazole,3-nitropyrazole,1h-pyrazole, 3-nitro,3-nitro-2h-pyrazole,nitropyrazole,pyrazole, 3-nitro,5-nitro-1h-pyrazol,djw 94,djw-94,5-nitropyrazole
123419
5-nitro-1H-pyrazole
C1=C(NN=C1)[N+](=O)[O-]
26621-44-3
C3H3N3O2
113.08
MZRUFMBFIKGOAL-UHFFFAOYSA-N
3-nitro-1h-pyrazole,3-nitropyrazole,1h-pyrazole, 3-nitro,3-nitro-2h-pyrazole,nitropyrazole,pyrazole, 3-nitro,5-nitro-1h-pyrazol,djw 94,djw-94,5-nitropyrazole
123419
5-nitro-1H-pyrazole
C1=C(NN=C1)[N+](=O)[O-]
Packaging Glass bottle
Quantity 5g
Melting Point 173.0°C to 175.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 113.08
Percent Purity 97%
Chemical Name or Material 3-Nitro-1H-pyrazole

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning