Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-2,6-lutidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
15513-52-7
C7H8N2O2
152.15
MFCD00023503
AETHUDGJSSKZKT-UHFFFAOYSA-N
3-nitro-2,6-lutidine,3-nitro-2,6-dimethylpyridine,pyridine, 2,6-dimethyl-3-nitro,nitrolutidine,pubchem14242,acmc-20a0s3,2,6-dimethyl-3-nitro-pyridine,pyridine,2,6-dimethyl-3-nitro,2-methyl-5-nitro-6-methylpyridine
84958
2,6-dimethyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])C
15513-52-7
C7H8N2O2
152.15
MFCD00023503
AETHUDGJSSKZKT-UHFFFAOYSA-N
3-nitro-2,6-lutidine,3-nitro-2,6-dimethylpyridine,pyridine, 2,6-dimethyl-3-nitro,nitrolutidine,pubchem14242,acmc-20a0s3,2,6-dimethyl-3-nitro-pyridine,pyridine,2,6-dimethyl-3-nitro,2-methyl-5-nitro-6-methylpyridine
84958
2,6-dimethyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])C
Packaging Glass bottle
Color Tan to White
Quantity 5g
Melting Point 32.0°C to 38.0°C
Flash Point 37.78°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Beilstein 20, V,6, 52
Formula Weight 152.15
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Nitro-2, 6-lutidine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 239-543-5