Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitro-4-(1-piperidinyl)benzaldehyde, 96%

Click to view available options
Quantity:
1g
5g
39911-29-0
C12H14N2O3
234.255
MFCD00831780
BNNMEAFYZHALEC-UHFFFAOYSA-N
3-nitro-4-piperidinobenzaldehyde,3-nitro-4-piperidin-1-yl benzaldehyde,3-nitro-4-piperidinobenzenecarbaldehyde,3-nitro-4-1-piperidinyl benzaldehyde,3-nitro-4-1-piperidino benzaldehyde,3-nitro-4-piperidylbenzaldehyde,acmc-20amzq,3-nitro-4-piperidin-1-yl-benzaldehyde,benzaldehyde, 3-nitro-4-1-piperidinyl
2737076
3-nitro-4-piperidin-1-ylbenzaldehyde
C1CCN(CC1)C2=C(C=C(C=C2)C=O)[N+](=O)[O-]
39911-29-0
C12H14N2O3
234.255
MFCD00831780
BNNMEAFYZHALEC-UHFFFAOYSA-N
3-nitro-4-piperidinobenzaldehyde,3-nitro-4-piperidin-1-yl benzaldehyde,3-nitro-4-piperidinobenzenecarbaldehyde,3-nitro-4-1-piperidinyl benzaldehyde,3-nitro-4-1-piperidino benzaldehyde,3-nitro-4-piperidylbenzaldehyde,acmc-20amzq,3-nitro-4-piperidin-1-yl-benzaldehyde,benzaldehyde, 3-nitro-4-1-piperidinyl
2737076
3-nitro-4-piperidin-1-ylbenzaldehyde
C1CCN(CC1)C2=C(C=C(C=C2)C=O)[N+](=O)[O-]
Sensitivity Air Sensitive
Quantity 5g
Melting Point 50°C to 54°C
Formula Weight 234.26
Percent Purity 96%
Chemical Name or Material 3-Nitro-4-(1-piperidinyl)benzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures