Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitro-5-(trifluoromethyl)benzoic acid, 97%

Click to view available options
Quantity:
5g
25g
328-80-3
C8H4F3NO4
235.118
MFCD00024509
ODCLHXGXGFBBTA-UHFFFAOYSA-N
3-nitro-5-trifluoromethyl benzoic acid,3-nitro-5-trifluoromethylbenzoic acid,3-trifluoromethyl-5-nitrobenzoic acid,3-carboxy-5-nitrobenzotrifluoride,3-nitro-5 trifluoromethyl benzoic acid,benzoic acid, 3-nitro-5-trifluoromethyl,pubchem4544,acmc-1cr4t,rarechem al bo 1012
616602
C1=C(C=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
328-80-3
C8H4F3NO4
235.118
MFCD00024509
ODCLHXGXGFBBTA-UHFFFAOYSA-N
3-nitro-5-trifluoromethyl benzoic acid,3-nitro-5-trifluoromethylbenzoic acid,3-trifluoromethyl-5-nitrobenzoic acid,3-carboxy-5-nitrobenzotrifluoride,3-nitro-5 trifluoromethyl benzoic acid,benzoic acid, 3-nitro-5-trifluoromethyl,pubchem4544,acmc-1cr4t,rarechem al bo 1012
616602
C1=C(C=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
Quantity 25g
Melting Point 127°C to 132°C
Beilstein 2284651
Formula Weight 235.12
Percent Purity 97%
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures