Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitro-alpha-toluenesulfonyl chloride, 95%

Click to view available options
Quantity:
1g
5g
25g
58032-84-1
C7H6ClNO4S
235.638
MFCD01859935
IUYFKLDRPZLAQQ-UHFFFAOYSA-N
3-nitrophenyl methanesulfonyl chloride,3-nitrobenzylsulfonyl chloride,3-nitrophenylmethanesulfonylchloride,benzenemethanesulfonyl chloride, 3-nitro,3-nitro-alpha-toluenesulfonyl chloride,chloro 3-nitrophenyl methyl sulfone,acmc-1b04d,3-nitro-phenylmethane-sulfonyl chloride,3-nitrophenyl methanesulphonyl chloride,3-nitrobenzenemethanesulfonyl chloride
13196775
(3-nitrophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)[N+](=O)[O-])CS(=O)(=O)Cl
58032-84-1
C7H6ClNO4S
235.638
MFCD01859935
IUYFKLDRPZLAQQ-UHFFFAOYSA-N
3-nitrophenyl methanesulfonyl chloride,3-nitrobenzylsulfonyl chloride,3-nitrophenylmethanesulfonylchloride,benzenemethanesulfonyl chloride, 3-nitro,3-nitro-alpha-toluenesulfonyl chloride,chloro 3-nitrophenyl methyl sulfone,acmc-1b04d,3-nitro-phenylmethane-sulfonyl chloride,3-nitrophenyl methanesulphonyl chloride,3-nitrobenzenemethanesulfonyl chloride
13196775
(3-nitrophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)[N+](=O)[O-])CS(=O)(=O)Cl
Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
Melting Point 95°C to 102°C
Quantity 25g
UN Number UN3261
Formula Weight 235.64
Percent Purity 95%
Chemical Name or Material 3-Nitro-alpha-toluenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.