Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Nitroacetanilide, 98+%

Click to view available options
Quantity:
5g
25g
100g
122-28-1
C8H8N2O3
180.163
MFCD00017015
KFTYNYHJHKCRKU-UHFFFAOYSA-N
n-3-nitrophenyl acetamide,3'-nitroacetanilide,m-nitroacetanilide,3-nitroacetanilide,acetamide, n-3-nitrophenyl,n-acetyl-m-nitroaniline,3-nitro-n-acetylaniline,acetanilide, 3'-nitro,unii-qgh8s22nbp,qgh8s22nbp
31206
N-(3-nitrophenyl)acetamide
CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]
122-28-1
C8H8N2O3
180.163
MFCD00017015
KFTYNYHJHKCRKU-UHFFFAOYSA-N
n-3-nitrophenyl acetamide,3'-nitroacetanilide,m-nitroacetanilide,3-nitroacetanilide,acetamide, n-3-nitrophenyl,n-acetyl-m-nitroaniline,3-nitro-n-acetylaniline,acetanilide, 3'-nitro,unii-qgh8s22nbp,qgh8s22nbp
31206
N-(3-nitrophenyl)acetamide
CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]
Quantity 5g
Melting Point 151°C to 153°C
Beilstein 2211961
Merck Index 14,6581
Formula Weight 180.16
Percent Purity ≥98%
Chemical Name or Material 3'-Nitroacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA